accessibility menu, dialog, popup

UserName

HClCH3OOCH3ONHONCH3CH3

CAS RN 122892-31-3

IUPAC Name: hydrogen N-({4-[2-(dimethylamino)ethoxy]phenyl}methyl)-3,4-dimethoxybenzamide chloride
Synonyms: hydrogen itopride chloride
Molecular Weight (g/mol): 394.9
Molecular Formula: C20H27ClN2O4
InChi Key: ZTOUXLLIPWWHSR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(C=C1OC)C(=O)NCC1=CC=C(OCCN(C)C)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)