accessibility menu, dialog, popup

UserName

H3COSFFFFFBr

CAS RN 1240257-53-7

IUPAC Name: 2-(bromomethyl)-1-methoxy-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 2-(bromomethyl)-1-methoxy-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 327.11
Molecular Formula: C8H8BrF5OS
InChi Key: VOLXPNDUPQNACF-UHFFFAOYSA-N
SMILES: COC1=C(CBr)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results