missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15477357
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477357 1 g 1g
15458925 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477357 Supplier Thermo Scientific Alfa Aesar Supplier No. H34415.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1240257-53-7
Molecular Formula C8H8BrF5OS
Molecular Weight (g/mol) 327.107
MDL Number MFCD16652488
InChI Key VOLXPNDUPQNACF-UHFFFAOYSA-N
Synonym 2-methoxy-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-methoxy-4-pentafluoro-??-sulfanyl benzene, 2-methoxy-5-pentafluorothio benzyl bromide
PubChem CID 66523611
IUPAC Name [3-(bromomethyl)-4-methoxyphenyl]-pentafluoro-$l^{6}-sulfane
SMILES COC1=C(C=C(C=C1)S(F)(F)(F)(F)F)CBr

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 327.11
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(pentafluorothio)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.