CAS RN 1241390-29-3
CAS RN 1241390-29-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ DFHBI
Mimic of GFP fluorophore for imaging RNA in living cells
2
3