accessibility menu, dialog, popup

UserName

CAS RN 1256482-94-6

CH3CH3H3COOHNONH2OH

CAS RN 1256482-94-6

IUPAC Name: tert-butyl N-(1-carbamoyl-3-hydroxypropan-2-yl)carbamate
Synonyms: tert-butyl N-(1-carbamoyl-3-hydroxypropan-2-yl)carbamate
Molecular Weight (g/mol): 218.25
Molecular Formula: C9H18N2O4
InChi Key: BYCKHNZSBNGBQL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CO)CC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)