CAS RN 125945-98-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzoic Acid-13C6, TRC
CAS: 125945-98-4 Molecular Formula: 13C6 C H6 O2 Molecular Weight (g/mol): 128.08 Synonym: Benzoic-1,2,3,4,5,6-13C6 acid,[Ring-U-13C]-benzoic acid,Benzoic Acid-13C6 IUPAC Name: (1,2,3,4,5,6-^{13}C_{6})cyclohexatrienecarboxylic acid SMILES: OC(=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1