Learn More
Benzoic Acid-13C6, TRC
Product Code. 30134999
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134999 250 mg 250mg
30010208 100 mg 100mg
30006813 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134999 Supplier Toronto Research Chemicals Supplier No. TRCB203903250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 125945-98-4
Molecular Formula 13C6 C H6 O2
Molecular Weight (g/mol) 128.08
Synonym Benzoic-1,2,3,4,5,6-13C6 acid, [Ring-U-13C]-benzoic acid, Benzoic Acid-13C6
IUPAC Name (1,2,3,4,5,6-^{13}C_{6})cyclohexatrienecarboxylic acid
SMILES OC(=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
Formula Weight 128.0569
Percent Purity >95%
Chemical Name or Material Benzoic acid ring-13C6
Grade HPLC