accessibility menu, dialog, popup

UserName

CAS RN 1273-98-9

Nd

CAS RN 1273-98-9

IUPAC Name: tris(cyclopenta-2,4-dien-1-yl)neodymium
Synonyms: tris(cyclopenta-2,4-dien-1-yl)neodymium
Molecular Weight (g/mol): 339.53
Molecular Formula: C15H15Nd
InChi Key: OACOCUZSZBRUKL-UHFFFAOYSA-N
SMILES: C1=CC(C=C1)[Nd](C1C=CC=C1)C1C=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)