accessibility menu, dialog, popup

UserName

CAS RN 127675-46-1

FOFCl

CAS RN 127675-46-1

IUPAC Name: 3-chloro-2,4-difluorobenzaldehyde
Synonyms: 3-chloro-2,4-difluorobenzaldehyde
Molecular Weight (g/mol): 176.55
Molecular Formula: C7H3ClF2O
InChi Key: PDBQQCWWJXDNJO-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C(F)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results