missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4-difluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 11431320
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431320 25 g 25g
11411320 1 g 1g
11421320 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431320 Supplier Thermo Scientific Alfa Aesar Supplier No. H26383.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2,4-difluorobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127675-46-1
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.547
MDL Number MFCD04116001
InChI Key PDBQQCWWJXDNJO-UHFFFAOYSA-N
PubChem CID 3735177
IUPAC Name 3-chloro-2,4-difluorobenzaldehyde
SMILES C1=CC(=C(C(=C1C=O)F)Cl)F

Specifications

Refractive Index 1.525
Quantity 25 g
Sensitivity Air Sensitive
Synonym 3-chloro-2,4-difluoro-benzaldehyde, benzaldehyde,3-chloro-2,4-difluoro, pubchem2886, acmc-20a7il, 3-chloro-2,4-difluorobenzaldehyde, benzaldehyde, 3-chloro-2,4-difluoro, 3-chloranyl-2,4-bis fluoranyl benzaldehyde
Solubility Information Slightly soluble in water.
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2,4-difluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.