accessibility menu, dialog, popup

UserName

NBrO

CAS RN 128071-75-0

IUPAC Name: 2-bromopyridine-3-carbaldehyde
Synonyms: 2-bromopyridine-3-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: GNFWMEFWZWXLIN-UHFFFAOYSA-N
SMILES: BrC1=C(C=O)C=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results