accessibility menu, dialog, popup

UserName

CAS RN 1302-93-8

OAlOAlOSiOO32

CAS RN 1302-93-8

IUPAC Name: tris(oxo[(oxoalumanyl)oxy]alumane); bis(silanedione)
Synonyms: tris(sapphire powder); bis(silica)
Molecular Weight (g/mol): 426.05
Molecular Formula: Al6O13Si2
InChi Key: KZHJGOXRZJKJNY-UHFFFAOYSA-N
SMILES: O=[Si]=O.O=[Si]=O.O=[Al]O[Al]=O.O=[Al]O[Al]=O.O=[Al]O[Al]=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results