accessibility menu, dialog, popup

UserName

CAS RN 1303-52-2

OH2AuOH2H2O

CAS RN 1303-52-2

IUPAC Name: gold(III) hydroxide
Synonyms: gold(III) hydroxide
Molecular Weight (g/mol): 251.01
Molecular Formula: AuH6O3
InChi Key: PPOYUERUQZXZBE-UHFFFAOYSA-N
SMILES: O.O.O.[Au]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results