accessibility menu, dialog, popup

UserName

CAS RN 13106-76-8

NH4NH4OMoOOO

CAS RN 13106-76-8

IUPAC Name: diammonium dioxomolybdenumbis(olate)
Synonyms: ammonium orthomolybdate
Molecular Weight (g/mol): 196.02
Molecular Formula: H8MoN2O4
InChi Key: APUPEJJSWDHEBO-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].[O-][Mo]([O-])(=O)=O
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)