accessibility menu, dialog, popup

UserName

CAS RN 1311-10-0

H2OH2OH2OH2OH2OH2OH2OH2OOHOHSr

CAS RN 1311-10-0

IUPAC Name: strontium(2+) octahydrate dihydroxide
Synonyms: strontium(2+) octahydrate dihydroxide
Molecular Weight (g/mol): 265.75
Molecular Formula: H18O10Sr
InChi Key: UJPWWRPNIRRCPJ-UHFFFAOYSA-L
SMILES: O.O.O.O.O.O.O.O.[OH-].[OH-].[Sr++]
Physical Form 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)