CAS RN 131674-41-4
IUPAC Name:
3-(2-methoxypyridin-3-yl)prop-2-enoic acid
Synonyms:
3-(2-methoxypyridin-3-yl)prop-2-enoic acid
Molecular Weight (g/mol):
179.18
Molecular Formula:
C9H9NO3
InChi Key:
MTVRDAQJMCORAQ-UHFFFAOYSA-N
SMILES:
COC1=C(C=CC(O)=O)C=CC=N1
1
–
1
of
1
results
1
3-(2-Methoxy-3-pyridyl)acrylic acid, 95%
CAS: 131674-41-4 Molecular Formula: C9H9NO3 Molecular Weight (g/mol): 179.175 MDL Number: MFCD07781156 InChI Key: MTVRDAQJMCORAQ-SNAWJCMRSA-N Synonym: 3-2-methoxy-3-pyridyl acrylic acid,e-3-2-methoxypyridin-3-yl acrylic acid,3-2-methoxypyridin-3-yl acrylic acid,2e-3-2-methoxypyridin-3-yl prop-2-enoic acid,2e-3-2-methoxy 3-pyridyl prop-2-enoic acid PubChem CID: 14691926 IUPAC Name: (E)-3-(2-methoxypyridin-3-yl)prop-2-enoic acid SMILES: COC1=C(C=CC=N1)C=CC(=O)O