accessibility menu, dialog, popup

UserName

CAS RN 131740-09-5

ClCH3N(R)(S)OHHOOHOOClH

CAS RN 131740-09-5

IUPAC Name: hydrogen 2-(2-chlorophenyl)-5,7-dihydroxy-8-[(3S,4R)-3-hydroxy-1-methylpiperidin-4-yl]-4H-chromen-4-one chloride
Synonyms: hydrogen flavopiridol chloride
Molecular Weight (g/mol): 438.3
Molecular Formula: C21H21Cl2NO5
InChi Key: LGMSNQNWOCSPIK-LWHGMNCYSA-N
SMILES: [H+].[Cl-].CN1CC[C@@H]([C@H](O)C1)C1=C2OC(=CC(=O)C2=C(O)C=C1O)C1=CC=CC=C1Cl
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results