accessibility menu, dialog, popup

UserName

H3COSOOClH3C

CAS RN 131880-55-2

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-6-sulfonyl chloride
Synonyms: 2,2-dimethyl-3,4-dihydro-1-benzopyran-6-sulfonyl chloride
Molecular Weight (g/mol): 260.73
Molecular Formula: C11H13ClO3S
InChi Key: UUQKIRXYJVLZAJ-UHFFFAOYSA-N
SMILES: CC1(C)CCC2=C(O1)C=CC(=C2)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results