accessibility menu, dialog, popup

UserName

CAS RN 133073-73-1

NNNNNHN(S)OOONH2NH2OHHOH3C

CAS RN 133073-73-1

IUPAC Name: 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl](methyl)amino}phenyl)formamido]pentanedioic acid dihydrate
Synonyms: dihydrate methotrexate
Molecular Weight (g/mol): 490.48
Molecular Formula: C20H26N8O7
InChi Key: TXQDMGIRZSHAQM-UHFFFAOYNA-N
SMILES: O.O.CN(CC1=CN=C2N=C(N)N=C(N)C2=N1)C1=CC=C(C=C1)C(=O)NC(CCC(O)=O)C(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results