accessibility menu, dialog, popup

UserName

CAS RN 13308-51-5

OPOBOO

CAS RN 13308-51-5

IUPAC Name: 2,4,5-trioxa-1λ⁵-phospha-3-borabicyclo[1.1.1]pentan-1-one
Synonyms: boron phosphate
Molecular Weight (g/mol): 105.78
Molecular Formula: BO4P
InChi Key: RKVCQBPDVHFKCU-UHFFFAOYSA-N
SMILES: O=P12OB(O1)O2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results