CAS RN 133454-55-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Iloperidone metabolite Hydroxy Iloperidone, MedChemExpress
MedChemExpress Iloperidone metabolite Hydroxy Iloperidone (P88; Hydroxy Iloperidone) is a metabolite of Iloperidone, which is an atypical antipsychotic.