accessibility menu, dialog, popup

UserName

CAS RN 134-64-5

CH3N(S)(R)O(S)HOCH3N(S)(R)O(S)HOOHSOHOO

CAS RN 134-64-5

IUPAC Name: bis(2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethan-1-one); sulfuric acid
Synonyms: bis((-)-lobeline); sulfuric acid
Molecular Weight (g/mol): 773
Molecular Formula: C44H56N2O8S
InChi Key: GRZMOSSVIPFGFF-GNJLJDPWSA-N
SMILES: OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results