CAS RN 13451-05-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
335.46
Molecular Formula:
O4SrW
InChi Key:
KXELQHMRKFESFN-UHFFFAOYSA-N
SMILES:
[Sr++].[O-][W]([O-])(=O)=O
1
–
1
of
1
results
1
Strontium tungsten oxide, 99.9% (metals basis)
CAS: 13451-05-3 MDL Number: MFCD00049562 Synonym: Strontium tungstate