accessibility menu, dialog, popup

UserName

CAS RN 13464-82-9

InInOSOOOOSOOOOSOOO

CAS RN 13464-82-9

IUPAC Name: diindium(3+) trisulfate
Synonyms: indium(III) sulfate
Molecular Weight (g/mol): 517.8
Molecular Formula: In2O12S3
InChi Key: XGCKLPDYTQRDTR-UHFFFAOYSA-H
SMILES: [In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)