CAS RN 1346601-65-7
CAS RN 1346601-65-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Deschloro Bupropion-d9 Hydrochloride, TRC
CAS: 1346601-65-7 Molecular Formula: C13H11D9ClNO Molecular Weight (g/mol): 250.81 Synonym: 2-[(1,1-Dimethylethyl)amino]-1-phenyl-1-propanone Hydrochloride,2-(tert-Butylamino)propiophenone Hydrochloride IUPAC Name: 2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-1-phenylpropan-1-one;hydrochloride SMILES: Cl.[2H]C([2H])([2H])C(NC(C)C(=O)c1ccccc1)(C([2H])([2H])[2H])C([2H])([2H])[2H]