missing translation for 'onlineSavingsMsg'
Learn More

Deschloro Bupropion-d9 Hydrochloride, TRC

Product Code. 30360346
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360346 2.5 mg 2.5mg
30358923 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30360346 Supplier Toronto Research Chemicals Supplier No. TRCD2885472.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346601-65-7
Molecular Formula C13H11D9ClNO
Molecular Weight (g/mol) 250.81
Synonym 2-[(1,1-Dimethylethyl)amino]-1-phenyl-1-propanone Hydrochloride, 2-(tert-Butylamino)propiophenone Hydrochloride
IUPAC Name 2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-1-phenylpropan-1-one;hydrochloride
SMILES Cl.[2H]C([2H])([2H])C(NC(C)C(=O)c1ccccc1)(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C13H19NO.ClH/c1-10(14-13(2,3)4)12(15)11-8-6-5-7-9-11;/h5-10,14H,1-4H3;1H/i2D3,3D3,4D3;
Formula Weight 250.1798
Chemical Name or Material Deschloro Bupropion-d9 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.