accessibility menu, dialog, popup

UserName

FH2NFOH

CAS RN 135086-76-9

IUPAC Name: 4-amino-3,5-difluorophenol
Synonyms: 4-amino-3,5-difluorophenol
Molecular Weight (g/mol): 145.11
Molecular Formula: C6H5F2NO
InChi Key: YHQVBHFKCBFNEG-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results