accessibility menu, dialog, popup

UserName

CAS RN 13510-35-5

InIII

CAS RN 13510-35-5

IUPAC Name: indium(3+) triiodide
Synonyms: indium(3+) triiodide
Molecular Weight (g/mol): 495.53
Molecular Formula: I3In
InChi Key: RMUKCGUDVKEQPL-UHFFFAOYSA-K
SMILES: [In+3].[I-].[I-].[I-]
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)