accessibility menu, dialog, popup

UserName

CAS RN 13520-72-4

HFBiHO

CAS RN 13520-72-4

IUPAC Name: fluorobismuthanone
Synonyms: fluorobismuthanone
Molecular Weight (g/mol): 245.99
Molecular Formula: BiFH2O
InChi Key: KHMIIGVSZMFUNW-UHFFFAOYSA-N
SMILES: F.O=[BiH]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results