CAS RN 1356020-01-3
CAS RN 1356020-01-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(4S)-2-[[[(2R)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]methyl]-5,5-dimethyl-4-thiazolidinecarboxylic Acid (Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.