accessibility menu, dialog, popup

UserName

CAS RN 13709-42-7

FFFNd

CAS RN 13709-42-7

IUPAC Name: neodymium(3+) trifluoride
Synonyms: neodymium(3+) trifluoride
Molecular Weight (g/mol): 201.24
Molecular Formula: F3Nd
InChi Key: XRADHEAKQRNYQQ-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Nd+3]
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)