accessibility menu, dialog, popup

UserName

CAS RN 13721-39-6

NaNaNaOVOOO

CAS RN 13721-39-6

IUPAC Name: trisodium oxovanadiumtris(olate)
Synonyms: sodium orthovanadate sodium vanadium oxide trisodium vanadate vanadic acid, trisodium salt sodium tetraoxovanadate(3-) Na3VO4 vanadic acid (H3VO4), sodium salt trisodium orthovanadate sodium vanadate(V)
Molecular Weight (g/mol): 183.91
Molecular Formula: Na3O4V
InChi Key: IHIXIJGXTJIKRB-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[Na+].[O-][V]([O-])([O-])=O
Molecular Weight (g/mol) 
  • (8)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (8)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)