CAS RN 1378863-74-1
CAS RN 1378863-74-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl Olmesartan Medoxomil, TRC
CAS: 1378863-74-1 Molecular Formula: C30 H32 N6 O6 Molecular Weight (g/mol): 572.61 Synonym: 1H-Imidazole-5-carboxylic acid, 4-(1-hydroxy-1-methylpropyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester IUPAC Name: (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 5-(2-hydroxybutan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate SMILES: CCCc1nc(c(C(=O)OCC2=C(C)OC(=O)O2)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)C(C)(O)CC