accessibility menu, dialog, popup

UserName

CAS RN 13803-39-9

OO

CAS RN 13803-39-9

IUPAC Name: 5-phenylfuran-2-carbaldehyde
Synonyms: 5-phenylfuran-2-carbaldehyde
Molecular Weight (g/mol): 172.18
Molecular Formula: C11H8O2
InChi Key: BMJHNNPEPBZULA-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(O1)C1=CC=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results