CAS RN 142-83-6
IUPAC Name:
(2E,4E)-hexa-2,4-dienal
Synonyms:
2,4-hexadienal sorbaldehyde sorbic aldehyde
Molecular Weight (g/mol):
96.13
Molecular Formula:
C6H8O
InChi Key:
BATOPAZDIZEVQF-MQQKCMAXSA-N
SMILES:
C\C=C\C=C\C=O
1
–
1
of
1
results
1
2,4-Hexadienal, predominantly trans,trans, 95%
CAS: 142-83-6 Molecular Formula: C6H8O Molecular Weight (g/mol): 96.129 MDL Number: MFCD00007004 InChI Key: BATOPAZDIZEVQF-MQQKCMAXSA-N Synonym: 2,4-hexadienal,hexa-2,4-dienal,sorbaldehyde,sorbic aldehyde,trans,trans-2,4-hexadienal,2e,4e-hexa-2,4-dienal,2,4-hexadienal, 2e,4e,2,4-hexadienal, e,e,2-propyleneacrolein,3-propyleneacrolein PubChem CID: 637564 ChEBI: CHEBI:82334 IUPAC Name: (2E,4E)-hexa-2,4-dienal SMILES: CC=CC=CC=O