CAS RN 142557-61-7
CAS RN 142557-61-7
IUPAC Name:
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-carboxamide
Synonyms:
7-amino-1-cyclopropyl-3-ethyl-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
Molecular Weight (g/mol):
289.3
Molecular Formula:
C13H15N5O3
InChi Key:
HWODCHXORCTEGU-UHFFFAOYSA-N
SMILES:
CCN1C(=O)N(C2CC2)C2=NC(N)=C(C=C2C1=O)C(N)=O