Learn More
Tocris Bioscience™ A 484954
GREENER_CHOICE
Product Code. 18084662
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18084662
Brand: Tocris Bioscience™ 4483/10
CaM kinase III (eEF-2 kinase) inhibitor
Inhibitor of eukaryotic elongation factor-2 (eEF-2) kinase (IC50 = 0.28 μM in enzymatic assay). Reduces eEF-2 phosphorylation with little effect on cancer cell growth.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 142557-61-7
Molecular Formula C13H15N5O3
Specifications
Quantity 10 mg
Formula Weight 289.29
Purity >98%
Molecular Weight (g/mol) 289.295
Chemical Name or Material A 484954