accessibility menu, dialog, popup

UserName

OOOOOOCHHCCHH3CH3CCH3H3CCH3H3CGa

CAS RN 14405-43-7

IUPAC Name: 4-({bis[(4-oxopent-2-en-2-yl)oxy]gallanyl}oxy)pent-3-en-2-one
Synonyms: gallium acetylacetonate
Molecular Weight (g/mol): 367.05
Molecular Formula: C15H21GaO6
InChi Key: ZVYYAYJIGYODSD-UHFFFAOYSA-K
SMILES: CC(=O)C=C(C)O[Ga](OC(C)=CC(C)=O)OC(C)=CC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results