accessibility menu, dialog, popup

UserName

CAS RN 1445-45-0

CH3OCH3OH3COH3C

CAS RN 1445-45-0

IUPAC Name: 1,1,1-trimethoxyethane
Synonyms: 1,1,1-trimethoxyethane
Molecular Weight (g/mol): 120.15
Molecular Formula: C5H12O3
InChi Key: HDPNBNXLBDFELL-UHFFFAOYSA-N
SMILES: COC(C)(OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)