CAS RN 144675-97-8
CAS RN 144675-97-8
IUPAC Name:
bis((2E)-but-2-enedioic acid); 6,9-bis[(2-aminoethyl)amino]-5H,10H-benzo[g]isoquinoline-5,10-dione
Synonyms:
bis(fumaric acid); pixantrone
Molecular Weight (g/mol):
557.52
Molecular Formula:
C25H27N5O10
InChi Key:
SVAGFBGXEWPNJC-LVEZLNDCSA-N
SMILES:
OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.NCCNC1=CC=C(NCCN)C2=C1C(=O)C1=CC=NC=C1C2=O
1
–
2
of
2
results
2
Pixantrone, MedChemExpress
MedChemExpress Pixantrone (BBR 2778) dimaleate is a topoisomerase II inhibitor and DNA intercalator, with anti-tumor activity.