accessibility menu, dialog, popup

UserName

CAS RN 14481-29-9

IUPAC Name: λ²-iron(2+) tetraammonium hexakis(iminomethanide)
Synonyms: λ²-iron(2+) tetraammonium hexakis(cyanide)
Molecular Weight (g/mol): 284.11
Molecular Formula: C6H16FeN10
InChi Key: ZXQVPEBHZMCRMC-UHFFFAOYSA-R
SMILES: [NH4+].[NH4+].[NH4+].[NH4+].[Fe++].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results