accessibility menu, dialog, popup

UserName

H3CNOH3CCH3(E/Z)(E/Z)

CAS RN 14575-25-8

IUPAC Name: 1-phenyl-4-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)but-2-en-1-one
Synonyms: 1-phenyl-4-(1,3,3-trimethylindol-2-ylidene)but-2-en-1-one
Molecular Weight (g/mol): 303.41
Molecular Formula: C21H21NO
InChi Key: FWAPKZOTGYVZSA-UHFFFAOYSA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C1=CC=CC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results