accessibility menu, dialog, popup

UserName

CH3OONNH2NCl

CAS RN 1458-03-3

IUPAC Name: methyl 3-amino-6-chloropyrazine-2-carboxylate
Synonyms: methyl 3-amino-6-chloropyrazine-2-carboxylate
Molecular Weight (g/mol): 187.58
Molecular Formula: C6H6ClN3O2
InChi Key: JGAJCAJHSHUPGH-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Cl)=CN=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results