accessibility menu, dialog, popup

UserName

CAS RN 14882-18-9

OOBiOH2O

CAS RN 14882-18-9

IUPAC Name: 4-oxo-2H,4H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate
Synonyms: 4-oxo-2H-benzo[d]1,3-dioxa-2-bismacyclohexan-2-yl hydrate bismuthi subsalicylas bismutum subsalicylicum bismuth oxide salicylate bismuth(III) subsalicylate bismuth bismuth oxysalicylate
Molecular Weight (g/mol): 363.1
Molecular Formula: C7H6BiO4
InChi Key: QBWLKDFBINPHFT-UHFFFAOYSA-L
SMILES: O.O=C1O[Bi]OC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results