accessibility menu, dialog, popup

UserName

CAS RN 149-74-6

CH3SiClCl

CAS RN 149-74-6

IUPAC Name: dichloro(methyl)phenylsilane
Synonyms: dichloromethylphenylsilane
Molecular Weight (g/mol): 191.13
Molecular Formula: C7H8Cl2Si
InChi Key: GNEPOXWQWFSSOU-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)