CAS RN 14923-17-2
CAS RN 14923-17-2
IUPAC Name:
N''-{4-[(diaminomethylidene)amino]butyl}guanidine; sulfuric acid
Synonyms:
N''-{4-[(diaminomethylidene)amino]butyl}guanidine; sulfuric acid
Molecular Weight (g/mol):
270.31
Molecular Formula:
C6H18N6O4S
InChi Key:
RWTGFMPOODRXIM-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.NC(N)=NCCCCN=C(N)N
1
–
2
of
2
results
1
Arcaine Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Arcaine Sulfate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.