CAS RN 15244-35-6
CAS RN 15244-35-6
IUPAC Name:
cadmium(2+) octahydrate sulfate
Synonyms:
cadmium(2+) octahydrate sulfate
Molecular Weight (g/mol):
352.59
Molecular Formula:
CdH16O12S
InChi Key:
XPGCBFBTBOFJAE-UHFFFAOYSA-L
SMILES:
O.O.O.O.O.O.O.O.[Cd++].[O-]S([O-])(=O)=O
1
–
2
of
2
results
1
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)
CAS: 15244-35-6 Molecular Formula: CdO4S Molecular Weight (g/mol): 208.47 MDL Number: MFCD00151655 InChI Key: QCUOBSQYDGUHHT-UHFFFAOYSA-L IUPAC Name: cadmium(2+) sulfate SMILES: [Cd++].[O-]S([O-])(=O)=O
2
Cadmium sulfate hydrate, 98%
CAS: 15244-35-6 Molecular Formula: CdO4S Molecular Weight (g/mol): 208.47 MDL Number: MFCD00151655 InChI Key: QCUOBSQYDGUHHT-UHFFFAOYSA-L IUPAC Name: cadmium(2+) sulfate SMILES: [Cd++].[O-]S([O-])(=O)=O