CAS RN 152684-53-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TLK117, MedChemExpress
MedChemExpress TLK117, the active metabolite of TLK199, selective inhibits Glutathione S-transferase P1-1 (GSTP1-1) with a Ki of 0.4 μM for GSTP. TLK117 also competitively inhibits glyoxalase I with a Ki of 0.56 μM.