CAS RN 153507-24-5
CAS RN 153507-24-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Iodide Iodate concentrate, For 1L standard solution, 0.005 M I2 (0.01N), Honeywell Fluka™
CAS: 153507-24-5 MDL Number: MFCD00011355