CAS RN 156-62-7
CAS RN 156-62-7
IUPAC Name:
calcium aminoformonitrile
Synonyms:
calcium cyanamide
Molecular Weight (g/mol):
82.12
Molecular Formula:
CH2CaN2
InChi Key:
STRTXDFFNXSZQB-UHFFFAOYSA-N
SMILES:
[Ca++].NC#N
1
–
1
of
1
results
1
Calcium cyanamide, tech.
CAS: 156-62-7 Molecular Formula: CCaN2 MDL Number: MFCD00064894